Learn More
Socorex™ Compact Dispenser 501 Model
Brand:  Socorex™ 501.0505
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10516912
  • 2795.00 DKK / Each
Estimated Shipment: 12-08-2024
to see stock.
Specifications
Compact Dispenser
0.4 to 5 mL
501
2 Years
Autoclavable
0.2 mL
500 mL