missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Combs for TV200 and TV400 Twin-Plate Vertical Gel Electrophoresis Unit

For use with TV200 Standard Twin-Plate Wide Format Mini-Gel and TV400 Standard Twin-Plate Maxi-Gel vertical electrophoresis units

Brand:  Fisherbrand™

Additional Details : Weight : 2.00000kg

 View more versions of this product

Product Code. 10756751

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Available in 0.75, 1, 1.5 and 2mm thickness
Specifications

Specifications

TV200 Standard Twin-Plate Wide Format Mini-Gel Electrophoresis Unit and TV400 Standard Twin-Plate Maxi-Gel Electrophoresis Unit
24
95 μL
Teeth: 20 mm
Comb
1 mm
4.75 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers