missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Combs for TV200 and TV400 Twin-Plate Vertical Gel Electrophoresis Unit

For use with TV200 Standard Twin-Plate Wide Format Mini-Gel and TV400 Standard Twin-Plate Maxi-Gel vertical electrophoresis units

525.00 DKK - 593.00 DKK

Specifications

No. of Wells 24
Width (Metric) Teeth 4.75 mm
View More Specs

Products 3
Product Code Brand Width (Metric) Teeth Volume (Metric) Well No. of Wells Thickness (Metric) Price Quantity & Availability  
Product Code Brand Width (Metric) Teeth Volume (Metric) Well No. of Wells Thickness (Metric) Price Quantity & Availability  
10448212
View Documents Promotion Details
Fisherbrand™
4.75 mm 8 μL 24 0.35 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10579591
View Documents Promotion Details
Fisherbrand™
4.75 mm 71 μL 24 0.75 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
10756751
View Documents Promotion Details
Fisherbrand™
4.75 mm 95 μL 24 1 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description

Description

  • Available in 0.75, 1, 1.5 and 2mm thickness
Specifications

Specifications

24
4.75 mm
Videos
SDS
Documents

Documents

Product Certifications