Learn More
Fisherbrand™ CO2 Backup Systems for Isotemp Chest Ultra-Low Temperature Freezers
Ensure optimal security for critical samples by using Fisherbrand™ Isotemp CO2 Backup Systems for Ultra-Low Temperature Freezers.
23130.00 DKK
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
17868010
|
Fisherbrand™
|
Isotemp 3 cu. ft. Chest Ultra-Low Temperature Freezer |
23130.00 DKK
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||
17808020
|
Fisherbrand™
|
Isotemp 13 and 20 cu. ft. Chest Ultra-Low Temperature Freezer |
23130.00 DKK
Each |
Estimated Shipment: 12-08-2024
Log in to see stock. |
|||||
Description
- Maintains temperature down to -67°C with CO2
Specifications
Factory Installed | |
-67°C |