Learn More
Fisherbrand™ CO2 Backup Systems for Isotemp Chest Ultra-Low Temperature Freezers
Ensure optimal security for critical samples by using Fisherbrand™ Isotemp CO2 Backup Systems for Ultra-Low Temperature Freezers.
23130.00 DKK
Specifications
Description Factory Installed
Temperature Range -67°C
View More Specs
Products 2
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
Product Code Brand For Use With (Equipment) Price Quantity & Availability  
17868010
View Documents Promotion Details
Fisherbrand™
Isotemp 3 cu. ft. Chest Ultra-Low Temperature Freezer
23130.00 DKK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 12-08-2024
to see stock.
Add to basket
17808020
View Documents Promotion Details
Fisherbrand™
Isotemp 13 and 20 cu. ft. Chest Ultra-Low Temperature Freezer
23130.00 DKK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 12-08-2024
to see stock.
Add to basket
Description
Description
  • Maintains temperature down to -67°C with CO2
Specifications
Specifications
Factory Installed
-67°C