Learn More
Fisherbrand™ CO2 Backup Systems for Isotemp Chest Ultra-Low Temperature Freezers
Ensure optimal security for critical samples by using Fisherbrand™ Isotemp CO2 Backup Systems for Ultra-Low Temperature Freezers.
Brand:  Fisherbrand™
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 17808020
  • 23130.00 DKK / Each
Estimated Shipment: 30-08-2024
to see stock.
Add to basket
Description
Description
  • Maintains temperature down to -67°C with CO2
Specifications
Specifications
Factory Installed
-67°C
Isotemp 13 and 20 cu. ft. Chest Ultra-Low Temperature Freezer