Learn More
Thermo Scientific™ CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers
Ensure optimal security for critical samples by using Thermo Scientific™ CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers.
Brand: Thermo Scientific™ FCO4567
Description
Ensure optimal security for critical samples by using Thermo Scientific™ CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers, for use with Thermo Scientific™ TSX, Forma™, HERAfreeze™, Revco™ and TS-Series freezers.
Features:- LN2 backup system maintains temperature down to -80°C with liquid nitrogen
- CO2 backup system maintains temperature down to -67°C with CO2
Ordering Information:
Available as field- or factory-installed. Models for 300 box capacity models are standalone. Field-installed units must be installed by qualified professionals.
Specifications
CO2 Backup, Field Installed | |
-80°C | |
TSX Series, Forma 89000, HERAfreeze HLE, Revco RLE, Revco TLE Series (400 box and above) | |
Boxes (300) |
Videos
-
Thermo Scientific Revco UxF freezers
-
Thermo Scientific Revco UxF freezers
Accessories and Services
-
Unity Lab Services™ Tech Direct Service Plan for Thermo Sci... Tech Direct plan for ULT Freezer & LN2 S... Product Code: 17200434
3755.44 DKK / Each -
Unity Lab Services™ Total Care Warranty for Thermo Scientif... Total Care Warranty for ULT Freezer & LN... Product Code: 17210414
3611.00 DKK / Each -
Unity Lab Services™ Total Care Service Plan for Thermo Scie... Total Care plan for ULT Freezer & LN2 St... Product Code: 17280444
10066.00 DKK / Each -
Unity Lab Services™ Extended Warranty Plan for Thermo Scien... Extended Warranty Service Plan for ULT F... Product Code: 17375080
7111.00 DKK / Each -
Unity Lab Services™ Cold Storage Equipment Requalification ... Requalification Service Cold Storage (fr... Product Code: 30178681
13650.00 DKK / 1 unit