Få mere at vide
Clopamide, TRC
Artikelnummer. 30137927
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137927 100 mg
17921285 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30137927 Leverandør Toronto Research Chemicals Leverandørnr. TRCC587160100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 636-54-4
Molekylær formel C14 H20 Cl N3 O3 S
Molekylvægt (g/mol) 345.84
Synonym Clopamide, 4-Chloro-N-[(2RS,6SR)-2,6-dimethylpiperidin-1-yl]-3-sulfamoylbenzamide, Benzamide, 3-(aminosulfonyl)-4-chloro-N-(2,6-dimethyl-1-piperidinyl)-, cis-, Benzamide, 4-chloro-N-(2,6-dimethylpiperidino)-3-sulfamoyl-, cis- (8CI), 4-Chloro-N-(cis-2,6-dimethylpiperidino)-3-sulfamoylbenzamide, Adurix, Aquex, Brinaldix, Chlosudimeprimyl, Clopamide, Clopamidum, DT 327, N-(cis-2',6'-Dimethyl-1'-piperidyl)-3-sulfamyl-4-chlorobenzamide
IUPAC navn 4-chloro-N-[(2R,6S)-2,6-dimethylpiperidin-1-yl]-3-sulfamoylbenzamide
SMIL C[C@@H]1CCC[C@H](C)N1NC(=O)c2ccc(Cl)c(c2)S(=O)(=O)N
Tekniske data
Kemisk navn eller materiale Clopamide
Mængde 100 mg
InChI formel InChI=1S/C14H20ClN3O3S/c1-9-4-3-5-10(2)18(9)17-14(19)11-6-7-12(15)13(8-11)22(16,20)21/h6-10H,3-5H2,1-2H3,(H,17,19)(H2,16,20,21)/t9-,10+
Formel vægt 345.0914