Få mere at vide
Tocris Bioscience™ Clobenpropit dihydrobromide
Highly potent H3 antagonist and H4 partial agonist
Brand: Tocris Bioscience™ 0752/50
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
An extremely potent histamine H3 antagonist/inverse agonist (pA2=9.93). Also displays partial agonist activity at H4 receptors; induces eosinophil shape change with an EC50 of 3 nM.Tekniske data
| N-(4-Chlorobenzyl)-S-[3-(4(5)-imidazolyl)propyl]isothiourea dihydrobromide | |
| >99% | |
| Soluble to 100 mM in water |
| 145231-35-2 | |
| 50 mg |
Produkttitel
Ser du en mulighed for forbedring?
For Research Use Only