CAS RN 145231-35-2
IUPAC Navn:
N'-[(4-chlorophenyl)methyl]{[3-(1H-imidazol-5-yl)propyl]sulfanyl}methanimidamide dihydrobromide
Synonymer:
clobenpropit dihydrobromide
Molekylvægt (g/mol):
470.65
Molekylformel:
C14H19Br2ClN4S
InChi Key:
JIJQPEZAVLJZBO-UHFFFAOYSA-N
SMILES:
Br.Br.NC(SCCCC1=CN=CN1)=NCC1=CC=C(Cl)C=C1
1
–
3
af
3
Resultater
1
Tocris Bioscience™ Clobenpropit dihydrobromide
Highly potent H3 antagonist and H4 partial agonist
2
3