Få mere at vide
Cleomiscosin A, TRC
Artikelnummer. 30349772
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30349772 1 mg
30363583 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30349772 Leverandør Toronto Research Chemicals Leverandørnr. TRCC5730501MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 76948-72-6
Molekylær formel C20H18O8
Molekylvægt (g/mol) 386.35
Synonym (2R,3R)-rel-2,3-Dihydro-3-(4-hydroxy-3-methoxyphenyl)-2-(hydroxymethyl)-5-methoxy-9H-pyrano[2,3-f]-1,4-benzodioxin-9-one, trans-(±)-2,3-Dihydro-3-(4-hydroxy-3-methoxyphenyl)-2-(hydroxymethyl)-5-methoxy-9H-pyrano[2,3-f]-1,4-benzodioxin-9-one, NSC 346197
IUPAC navn (2R,3R)-3-(4-hydroxy-3-methoxyphenyl)-2-(hydroxymethyl)-5-methoxy-2,3-dihydro-9H-pyrano[2',3':3,4]benzo[1,2-b][1,4]dioxin-9-one
SMIL OC(C(OC)=C1)=CC=C1[C@H]([C@@H](CO)O2)OC3=C2C(OC(C=C4)=O)=C4C=C3OC
Tekniske data
Kemisk navn eller materiale Cleomiscosin A
Mængde 1 mg
InChI formel InChI=1S/C20H18O8/c1-24-13-7-10(3-5-12(13)22)17-15(9-21)26-20-18-11(4-6-16(23)27-18)8-14(25-2)19(20)28-17/h3-8,15,17,21-22H,9H2,1-2H3/t15-,17-/m1/s1
Formel vægt 386.1