Learn More
Heidolph™ Clamp for Erlenmeyer Flasks
Clamps for use with various perforated platforms
Brand: Heidolph™ 549-55000-00
This item is not returnable.
View return policy
Description
- Maximum number of clamps per platform type:
- 549-51000-00
- For platform 100 - max. 16
- For platform 1000 - max. 20
- For platform 2000 - max. 36
- 549-52000-00
- For platform 100 - max. 16
- For platform 1000 - max. 20
- For platform 2000 - max. 36
- 549-55000-00
- For platform 100 - max. 3
- For platform 1000 - max. 4
- For platform 2000 - max. 9
- 549-56000-00
- For platform 100 - max. 2
- For platform 1000 - max. 4
- For platform 2000 - max. 5
- 549-63000-00
- For platform 1000 - max. 2
- For platform 2000 - max. 3
- 549-51000-00
Specifications
500 mL Erlenmeyer flasks | |
Clamp | |
Perforated platforms 100, 1000 and 2000 | |
5 |
Product Content Correction
Product Title
Heidolph™ Clamp for Erlenmeyer Flasks > Clamp - Size 5 For 500 mL Erlenmeyer flasks
Spot an opportunity for improvement?Share a Content Correction