Learn More
Fisherbrand™ Cellulose Stoppers
For flasks, erlenmeyers and test tubes.
Brand: Fisherbrand™ 0005N00062
This item is not returnable.
View return policy
Description
- Permeable in air
- Sterilizable to 200°C maximum
- Single use
Specifications
Single-use | |
32 mm | |
200°C | |
18 mm | |
Stopper |
Beakers, Erlenmeyer flasks, tubes, and different types of bottles | |
1000PK | |
13 mm | |
Cellulose Fiber | |
Sterile |
Product Content Correction
Product Title
Fisherbrand™ Cellulose Stoppers > Size: 13 x 18 x 32mm
Spot an opportunity for improvement?Share a Content Correction