Learn More
Fisherbrand™ Cellulose Stoppers
For flasks, erlenmeyers and test tubes.
Brand: Fisherbrand™ 0005N00080
This item is not returnable.
View return policy
Description
- Permeable in air
- Sterilizable to 200°C maximum
- Single use
Specifications
Single-use | |
60 mm | |
200°C | |
46 mm | |
Stopper |
Beakers, Erlenmeyer flasks, tubes, and different types of bottles | |
100PK | |
35 mm | |
Cellulose Fiber | |
Sterile |
Product Content Correction
Product Title
Fisherbrand™ Cellulose Stoppers > Size: 35 x 46 x 60mm
Spot an opportunity for improvement?Share a Content Correction