Få mere at vide
CJ-42794, MedChemExpress
Artikelnummer. 16455126
Encompass_Preferred
Skift visning
Mængde:
10 mg
50 mg
100 mg
Pakningsstørrelse:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16455126 100 mg -
16414168 10 mg -
16410517 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 16455126 Leverandør Medchem Express Leverandørnr. HY10797
MedChemExpress CJ-42794 is a selective prostaglandin E receptor subtype 4 (EP4) antagonist, inhibits [3H]-PGE2 binding to the human EP4 receptor with a mean pKi of 8.5, a binding affinity that was at least 200-fold more selective for the human EP4 receptor than other human EP receptor subtypes (EP1, EP2, and EP3).
CJ-42794 is a selective prostaglandin E receptor subtype 4
Tekniske data
Kemisk navn eller materiale CJ-42794
CAS 847728-01-2
Farve Off-White
Renhedsgrad noter Research
Molekylær formel C22H17ClFNO4
Mængde 100 mg
Synonym CJ-042794
Opløselighedsinformation DMSO : ≥ 28 mg/mL (67.66 mM)
SMIL ClC1=CC=C(OC2=CC=C(F)C=C2)C(C(N[C@H](C3=CC=C(C(O)=O)C=C3)C)=O)=C1
Molekylvægt (g/mol) 413.83
Formel vægt 413.83
Procent renhed 98.0%
Grad Research
Fysisk form Solid
Til brug med (applikation) COVID-19-immunoregulation