Få mere at vide
Citric acid-2,4-13C2, TRC
Artikelnummer. 30135530
Skift visning
Mængde:
25 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135530 50 mg
30137073 100 mg
30004018 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30135530 Leverandør Toronto Research Chemicals Leverandørnr. TRCC52106050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 121633-50-9
Molekylær formel 13C2 C4 H8 O7
Molekylvægt (g/mol) 194.11
Synonym 2-Hydroxy-1,2,3-propane-1,3-13C2-tricarboxylic Acid
IUPAC navn 2-hydroxy(1,3-^{13}C_{2})propane-1,2,3-tricarboxylic acid
SMIL OC(=O)[13CH2]C(O)([13CH2]C(=O)O)C(=O)O
Tekniske data
Kemisk navn eller materiale Citric acid-2,4-13C2
Mængde 50 mg
InChI formel InChI=1S/C6H8O7/c7-3(8)1-6(13,5(11)12)2-4(9)10/h13H,1-2H2,(H,7,8)(H,9,10)(H,11,12)/i1+1,2+1
Formel vægt 194.0337