Få mere at vide
cis-Vitamin K1, TRC
Artikelnummer. 30138553
Skift visning
Mængde:
1 mg
2 mg
5 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30138553 2 mg
30011216 1 mg
30007246 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30138553 Leverandør Toronto Research Chemicals Leverandørnr. TRCV6760702MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 16033-41-3
Molekylær formel C31 H46 O2
Molekylvægt (g/mol) 450.7
Synonym cis-Phytomenadione, 1,4-Naphthalenedione, 2-methyl-3-[(2Z,7R,11R)-3,7,11,15-tetramethyl-2-hexadecen-1-yl]-, 1,4-Naphthalenedione, 2-methyl-3-(3,7,11,15-tetramethyl-2-hexadecenyl)-, [R-[R*,R*-(Z)]]-, 1,4-Naphthalenedione, 2-methyl-3-[(2Z,7R,11R)-3,7,11,15-tetramethyl-2-hexadecenyl]- (9CI), Phylloquinone, cis- (8CI), cis-Phylloquinone, cis-Vitamin K
IUPAC navn 2-methyl-3-[(Z,7R,11R)-3,7,11,15-tetramethylhexadec-2-enyl]naphthalene-1,4-dione
SMIL CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC\C(=C/CC1=C(C)C(=O)c2ccccc2C1=O)\C
Tekniske data
Kemisk navn eller materiale cis-Vitamin K1
Renhedsgrad noter HPLC
Mængde 2 mg
InChI formel InChI=1S/C31H46O2/c1-22(2)12-9-13-23(3)14-10-15-24(4)16-11-17-25(5)20-21-27-26(6)30(32)28-18-7-8-19-29(28)31(27)33/h7-8,18-20,22-24H,9-17,21H2,1-6H3/b25-20-/t23-,24-/m1/s1
Formel vægt 450.3498
Procent renhed >95%