Få mere at vide
cis/trans-Haloperidol N-Oxide, TRC
Artikelnummer. 30007888
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30007888 5 mg
30005975 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30007888 Leverandør Toronto Research Chemicals Leverandørnr. TRCH1037355MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 148406-51-3
Molekylær formel C21H23ClFNO3
Molekylvægt (g/mol) 391.86
Synonym 4-[-4-(4-Chlorophenyl)-4-hydroxy-1-oxido-1-piperidinyl]-1-(4-fluorophenyl)-1-butanone, 4-[4-(4-Chlorophenyl)-4-hydroxy-1-oxido-1-piperidinyl]-1-(4-fluorophenyl)-1-butanone, 4-[4-(4-Chlorophenyl)-4-hydroxy-1-piperidinyl]-1-(4-fluorophenyl)-1-butanone N-Oxide
IUPAC navn 4-[4-(4-chlorophenyl)-4-hydroxy-1-oxidopiperidin-1-ium-1-yl]-1-(4-fluorophenyl)butan-1-one
SMIL OC1(CC[N+]([O-])(CCCC(=O)c2ccc(F)cc2)CC1)c3ccc(Cl)cc3
Tekniske data
Kemisk navn eller materiale cis/trans-Haloperidol N-Oxide
Mængde 5 mg
InChI formel InChI=1S/C21H23ClFNO3/c22-18-7-5-17(6-8-18)21(26)11-14-24(27,15-12-21)13-1-2-20(25)16-3-9-19(23)10-4-16/h3-10,26H,1-2,11-15H2
Formel vægt 391.135