Få mere at vide
cis-Tranexamic Acid, TRC
Artikelnummer. 30008266
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30008266 100 mg
30011669 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30008266 Leverandør Toronto Research Chemicals Leverandørnr. TRCT714500100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1197-17-7
Molekylær formel C8 H15 N O2
Molekylvægt (g/mol) 157.21
Synonym Cyclohexanecarboxylic acid, 4-(aminomethyl)-, cis-, cis-4-(Aminomethyl)cyclohexanecarboxylic acid, cis-4-(Aminomethyl)-1-cyclohexanecarboxylic acid, cis-AMCHA, cis-Tranexamic acid, cis-4-(Aminomethyl)cyclohexanecarboxylic Acid, cis-Tranexamic Acid, Tranexamic acid impurity B CRS
IUPAC navn 4-(aminomethyl)cyclohexanecarboxylic acid
SMIL NC[C@@H]1CC[C@@H](CC1)C(=O)O
Tekniske data
Kemisk navn eller materiale cis-Tranexamic Acid
Renhedsgrad noter HPLC
Mængde 100 mg
InChI formel InChI=1S/C8H15NO2/c9-5-6-1-3-7(4-2-6)8(10)11/h6-7H,1-5,9H2,(H,10,11)/t6-,7+
Formel vægt 157.1103
Procent renhed >95%