Få mere at vide
cis-Propenylphosphonic Acid (R)-(+)-Alpha-Methylbenzylamine Salt, TRC
Artikelnummer. 30135182
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135182 500 mg
30004172 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30135182 Leverandør Toronto Research Chemicals Leverandørnr. TRCP768980500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 933443-26-6
Molekylær formel C3H7O3P(C8H11N)
Molekylvægt (g/mol) 243.24
Synonym P-(1Z)-1-Propen-1-ylphosphonic Acid (R)-(+)-alpha-Methylbenzylamine Salt, (1Z)-1-Propenylphosphonic Acid (R)-(+)-alpha-Methylbenzylamine Salt, (Z)-1-Propenylphosphonic Acid (R)-(+)-alpha-Methylbenzylamine Salt, (Z)-Propenylphosphonic Acid (R)-(+)-alpha-Methylbenzylamine Salt, cis-1-Propenyl)phosphonic Acid (R)-(+)-alpha-Methylbenzylamine Salt, [(Z)-1-Propenyl]phosphonic Acid (R)-(+)-alpha-Methylbenzylamine Salt
IUPAC navn (1R)-1-phenylethanamine;[(Z)-prop-1-enyl]phosphonic acid
SMIL C\C=C/P(=O)(O)O.C[C@@H](N)c1ccccc1
Tekniske data
Kemisk navn eller materiale cis-Propenylphosphonic Acid (R)-(+)-alpha-Methylbenzylamine Salt
Mængde 500 mg
InChI formel InChI=1S/C8H11N.C3H7O3P/c1-7(9)8-5-3-2-4-6-8;1-2-3-7(4,5)6/h2-7H,9H2,1H3;2-3H,1H3,(H2,4,5,6)/b;3-2-/t7-;/m1./s1
Formel vægt 243.1024