CAS RN 933443-26-6
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
1
af
1
Resultater
1
cis-Propenylphosphonic Acid (R)-(+)-Alpha-Methylbenzylamine Salt, TRC
CAS: 933443-26-6 Molekylær formel: C3H7O3P(C8H11N) Molekylvægt (g/mol): 243.24 Synonym: P-(1Z)-1-Propen-1-ylphosphonic Acid (R)-(+)-alpha-Methylbenzylamine Salt,(1Z)-1-Propenylphosphonic Acid (R)-(+)-alpha-Methylbenzylamine Salt,(Z)-1-Propenylphosphonic Acid (R)-(+)-alpha-Methylbenzylamine Salt,(Z)-Propenylphosphonic Acid (R)-(+)-alpha-Methylbenzylamine Salt,cis-1-Propenyl)phosphonic Acid (R)-(+)-alpha-Methylbenzylamine Salt,[(Z)-1-Propenyl]phosphonic Acid (R)-(+)-alpha-Methylbenzylamine Salt IUPAC navn: (1R)-1-phenylethanamine;[(Z)-prop-1-enyl]phosphonic acid SMIL: C\C=C/P(=O)(O)O.C[C@@H](N)c1ccccc1