Få mere at vide
Cinchonidine Dihydrochloride, TRC
Artikelnummer. 30363769
Skift visning
Mængde:
250 mg
500 mg
2.5 g
Pakningsstørrelse:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30363769 500 mg -
30360286 2.5 g 2.5g
30351030 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30363769 Leverandør Toronto Research Chemicals Leverandørnr. TRCC475868500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 524-54-9
Molekylær formel C19H22N2O . 2HCl
Molekylvægt (g/mol) 367.3
IUPAC navn (R)-[(2S,4S,5R)-5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl]-quinolin-4-ylmethanol;dihydrochloride
SMIL Cl.Cl.O[C@@H]([C@@H]1C[C@@H]2CCN1C[C@@H]2C=C)c3ccnc4ccccc34
Tekniske data
Kemisk navn eller materiale Cinchonidine Dihydrochloride
Mængde 500 mg
InChI formel InChI=1S/C19H22N2O.2ClH/c1-2-13-12-21-10-8-14(13)11-18(21)19(22)16-7-9-20-17-6-4-3-5-15(16)17;;/h2-7,9,13-14,18-19,22H,1,8,10-12H2;2*1H/t13-,14-,18-,19+;;/m0../s1
Formel vægt 366.1266