Få mere at vide
CID5721353, MedChemExpress
Artikelnummer. 16483048
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
25 mg
50 mg
100 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16483048 10 mg
16455787 5 mg
16439616 50 mg
16422596 25 mg
16404356 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Artikelnummer. 16483048 Leverandør Medchem Express Leverandørnr. HY100502
MedChemExpress CID5721353 is an inhibitor of BCL6 with an IC50 value of 212 μM, which corresponds to a Ki of 147 μM.
CID5721353 is an inhibitor of BCL6 with an IC50 value of 212 μM, which corresponds to a Ki of 147 μM.
Tekniske data
Kemisk navn eller materiale CID5721353
CAS 301356-95-6
Farve Brown
Renhedsgrad noter Research
Molekylær formel C15H9BrN2O6S2
Mængde 10 mg
Opløselighedsinformation DMSO : 50 mg/mL (109.34 mM; Need ultrasonic)
SMIL O=C(O)C(N(C/1=O)C(SC1=C2C(NC3=C\2C=C(Br)C=C3)=O)=S)CC(O)=O
Molekylvægt (g/mol) 457.28
Formel vægt 457.28
Procent renhed 98.0%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-programmed cell death