Learn More
Macherey-Nagel™ Chromafix™ C18 Cartridge
Brand: Macherey-Nagel™ 731802
Additional Details : Weight : 0.01000kg
Specifications
Silica | |
480 mg | |
45 μm | |
Chromafix C18 Cartridge | |
50 Pack |
14% | |
Non-Polar Compounds, Pesticides | |
C18 | |
60 Å | |
500 m2/g |