Learn More
Macherey-Nagel™ Chromabond™ Florisil Polypropylene SPE Column
Brand: Macherey-Nagel™ 730082
This item is not returnable.
View return policy
Specifications
6 mL | |
1 g | |
Solid Phase Extraction Column |
SPE | |
150 to 250 ÎĽm |
Product Content Correction
Product Title
Macherey-Nagel™ Chromabond™ Florisil Polypropylene SPE Column > Column Capacity: 6mL; Adsorbent Weight: 1g
Spot an opportunity for improvement?Share a Content Correction