missing translation for 'onlineSavingsMsg'
Learn More
Cholesterol, ≥99%, MP Biomedicals™ Product Code.: 11493310

Cholesterol, ≥99%, MP Biomedicals™

Product Code. p-4404332 Shop All MP Biomedical Products
5 g, 5g
Click to view available options
Quantity:
5 g
100 g
Unit Size:
100g
5g
This item is not returnable. View return policy

Product Code. 11493310

Brand: MP Biomedicals 0210138205

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 57-88-5
Molecular Formula C27H46O
Molecular Weight (g/mol) 386.664
MDL Number MFCD00003646
InChI Key HVYWMOMLDIMFJA-DPAQBDIFSA-N
Synonym cholesterol, cholesterin, cholest-5-en-3beta-ol, cholesteryl alcohol, cholestrin, cordulan, dusoline, dusoran, provitamin d, cholesterine
PubChem CID 5997
ChEBI CHEBI:16113
IUPAC Name (3S,8S,9S,10R,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-3-ol
SMILES CC(C)CCCC(C)C1CCC2C1(CCC3C2CC=C4C3(CCC(C4)O)C)C

Specifications

Color White
Density 1.067g/mL
Assay Percent Range ≥99%
Quantity 5 g
Formula Weight 386.64
Physical Form Powder
Chemical Name or Material Cholesterol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cholesterol, ≥99%, MP Biomedicals™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.