Få mere at vide
Chlorpyriphos-methyl-d6, TRC
Artikelnummer. 30363711
Skift visning
Mængde:
0.5 mg
2.5 mg
Pakningsstørrelse:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30363711 2.5 mg 2.5mg
30360331 0.5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30363711 Leverandør Toronto Research Chemicals Leverandørnr. TRCC4253322.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2083629-84-7
Molekylær formel C7 2H6 H Cl3 N O3 P S
Molekylvægt (g/mol) 328.57
Synonym Chlorpyrifos-methyl D6, sulfanylidene-(3,5,6-trichloropyridin-2-yl)oxy-bis(trideuteriomethoxy)-λ^{5}-phosphane
IUPAC navn sulfanylidene-(3,5,6-trichloropyridin-2-yl)oxy-bis(trideuteriomethoxy)-λ^{5}-phosphane
SMIL [2H]C([2H])([2H])OP(=S)(Oc1nc(Cl)c(Cl)cc1Cl)OC([2H])([2H])[2H]
Tekniske data
Kemisk navn eller materiale Chlorpyrifos-methyl D6
Mængde 2.5 mg
InChI formel InChI=1S/C7H7Cl3NO3PS/c1-12-15(16,13-2)14-7-5(9)3-4(8)6(10)11-7/h3H,1-2H3/i1D3,2D3
Formel vægt 326.9326