missing translation for 'onlineSavingsMsg'
Få mere at vide

CGS 21680 hydrochloride, 99%, Thermo Scientific™

An A2A Adenosine receptor agonist

2820.94 DKK

Kemiske identifikatorer

CAS 120225-54-9
Molekylær formel C23H30ClN7O6
Molekylvægt (g/mol) 535.986
MDL nummer MFCD11045878
InChI nøgle QPHVMNOEKKJYJO-XWOZMEDYSA-N
Synonym cgs-21680 hydrochloride hydrate, 3-4-2-6-amino-9-2r,3r,4s,5s-5-ethylcarbamoyl-3,4-dihydroxyoxolan-2-yl purin-2-yl amino ethyl phenyl propanoic acid hydrate hydrochloride, c23h29n7o6.clh.h2o, cgs-21680 hydrochloride hydrate, solid, 2-p-2-carboxyethyl phenethylamino-5 inverted exclamation marka-n-ethylcarboxamidoadenosine hydrochloride hydrate
PubChem CID 133639586
IUPAC navn 3-[4-[2-[[6-amino-9-[(2S,4S,5R)-5-(ethylcarbamoyl)-3,4-dihydroxyoxolan-2-yl]purin-2-yl]amino]ethyl]phenyl]propanoic acid;hydrochloride
SMIL CCNC(=O)C1C(C(C(O1)N2C=NC3=C2N=C(N=C3N)NCCC4=CC=C(C=C4)CCC(=O)O)O)O.Cl
Se flere specifikationer

Produkter 1
Produktkode Brand Mængde Pris Antal & tilgængelighed  
Produktkode Brand Mængde Pris Antal & tilgængelighed  
15473499
Dokumentvisning
Thermo Scientific Alfa Aesar
J60793.MA
10mg
2820.94 DKK
10mg
Se alternative produkter
Spar op til 
»
Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!
Beskrivelse

Beskrivelse

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
CGS 21680 hydrochloride is currently used in research in studying neuronal transmission, also in respiration. CGS 21680 is commonly used to study the actions of the adenosine A2A receptor in cells and tissues. CGS 21680 HCl is an adenosine A2 receptor agonist with IC50 of 22 nM, exhibits 140-fold over A1 receptor. CGS 21680C effectively increases coronary flow with an ED25 value of 1.8 nM. In hippocampal slices, CGS 21680 apis weak agonist on pre- and postsynaptic measures of electrophysiologicaJ activity.

Solubility
Soluble up to 3.4 mg/mL in DMSO and 20 mg/mL in 45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Tekniske data

Kemiske identifikatorer

120225-54-9
535.986
QPHVMNOEKKJYJO-XWOZMEDYSA-N
133639586
CCNC(=O)C1C(C(C(O1)N2C=NC3=C2N=C(N=C3N)NCCC4=CC=C(C=C4)CCC(=O)O)O)O.Cl
C23H30ClN7O6
MFCD11045878
cgs-21680 hydrochloride hydrate, 3-4-2-6-amino-9-2r,3r,4s,5s-5-ethylcarbamoyl-3,4-dihydroxyoxolan-2-yl purin-2-yl amino ethyl phenyl propanoic acid hydrate hydrochloride, c23h29n7o6.clh.h2o, cgs-21680 hydrochloride hydrate, solid, 2-p-2-carboxyethyl phenethylamino-5 inverted exclamation marka-n-ethylcarboxamidoadenosine hydrochloride hydrate
3-[4-[2-[[6-amino-9-[(2S,4S,5R)-5-(ethylcarbamoyl)-3,4-dihydroxyoxolan-2-yl]purin-2-yl]amino]ethyl]phenyl]propanoic acid;hydrochloride

Tekniske data

CGS 21680 hydrochloride
C23H30ClN7O6
cgs-21680 hydrochloride hydrate, 3-4-2-6-amino-9-2r,3r,4s,5s-5-ethylcarbamoyl-3,4-dihydroxyoxolan-2-yl purin-2-yl amino ethyl phenyl propanoic acid hydrate hydrochloride, c23h29n7o6.clh.h2o, cgs-21680 hydrochloride hydrate, solid, 2-p-2-carboxyethyl phenethylamino-5 inverted exclamation marka-n-ethylcarboxamidoadenosine hydrochloride hydrate
QPHVMNOEKKJYJO-XWOZMEDYSA-N
3-[4-[2-[[6-amino-9-[(2S,4S,5R)-5-(ethylcarbamoyl)-3,4-dihydroxyoxolan-2-yl]purin-2-yl]amino]ethyl]phenyl]propanoic acid;hydrochloride
133639586
99%
120225-54-9
MFCD11045878
Soluble up to 3.4mg/mL in DMSO and 20mg/mL in 45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin.
CCNC(=O)C1C(C(C(O1)N2C=NC3=C2N=C(N=C3N)NCCC4=CC=C(C=C4)CCC(=O)O)O)O.Cl
535.986
535.98
Videoer
Sikkerhed og håndtering

Sikkerhed og håndtering

missing translation for 'tsca' : No

Anbefalet opbevaring : Ambient temperatures

SDS
Dokumenter

Dokumenter

Produktcertifikater

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.