Få mere at vide
Cetirizine Amide, TRC
Artikelnummer. 30133099
Skift visning
Mængde:
10 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133099 100 mg
30133079 10 mg
30007397 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133099 Leverandør Toronto Research Chemicals Leverandørnr. TRCC291080100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 83881-37-2
Molekylær formel C21 H26 Cl N3 O2
Molekylvægt (g/mol) 387.9
Synonym Acetamide, 2-[2-[4-[(4-chlorophenyl)phenylmethyl]-1-piperazinyl]ethoxy]-, 2-[2-[4-[(4-Chlorophenyl)phenylmethyl]-1-piperazinyl]ethoxy]acetamide, UCB-L 172, Cetirizine Amide
IUPAC navn 2-[2-[4-[(4-chlorophenyl)-phenylmethyl]piperazin-1-yl]ethoxy]acetamide
SMIL NC(=O)COCCN1CCN(CC1)C(c2ccccc2)c3ccc(Cl)cc3
Tekniske data
Kemisk navn eller materiale Cetirizine Amide
Mængde 100 mg
InChI formel InChI=1S/C21H26ClN3O2/c22-19-8-6-18(7-9-19)21(17-4-2-1-3-5-17)25-12-10-24(11-13-25)14-15-27-16-20(23)26/h1-9,21H,10-16H2,(H2,23,26)
Formel vægt 387.1714