Få mere at vide
Centrophenoxine hydrochloride, 98%, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar J64196.14
| Mængde | 25g |
|---|
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Ester of dimethylethanolamine and 4-chlorophenoxyacetic acid; activates acetylcholinesterase activity in hippocampus of aged rats
Kemiske identifikatorer
| C12H17Cl2NO3 | |
| MFCD00012533 | |
| meclofenoxate hydrochloride, centrophenoxine hydrochloride, lucidril, cerutil, meclofenoxate hcl, atsefen, helfergin, marucotol, acefen, brenal | |
| hydrogen 2-(dimethylamino)ethyl 2-(4-chlorophenoxy)acetate chloride |
Tekniske data
| Centrophenoxine hydrochloride | |
| C12H17Cl2NO3 | |
| 25g | |
| meclofenoxate hydrochloride, centrophenoxine hydrochloride, lucidril, cerutil, meclofenoxate hcl, atsefen, helfergin, marucotol, acefen, brenal | |
| [H+].[Cl-].CN(C)CCOC(=O)COC1=CC=C(Cl)C=C1 | |
| 294.17 | |
| 294.17 |
| 3685-84-5 | |
| MFCD00012533 | |
| 14,5780 | |
| FIVHOHCAXWQPGC-UHFFFAOYSA-N | |
| hydrogen 2-(dimethylamino)ethyl 2-(4-chlorophenoxy)acetate chloride | |
| 19379 | |
| 98% |