Få mere at vide
Centrinone-B, MedChemExpress
Artikelnummer. 16497578
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
Pakningsstørrelse:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16497578 10 mg 10mg
16493597 50 mg -
16450058 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 16497578 Leverandør Medchem Express Leverandørnr. HY18683
MedChemExpress Centrinone-B (LCR-323) is a potent and highly selective PLK4 inhibitor, with a Ki of 0.59 nM.
Centrinone-B (LCR-323) is a potent and highly selective PLK4 inhibitor, with a Ki of 0.59 nM.
Tekniske data
Kemisk navn eller materiale Centrinone-B
CAS 1798871-31-4
Farve Yellow
Renhedsgrad noter Research
Molekylær formel C27H27F2N7O5S2
Mængde 10 mg
Synonym LCR-323
Opløselighedsinformation DMSO : 25 mg/mL (39.58 mM; Need ultrasonic)
SMIL FC(C([N+]([O-])=O)=CC=C1)=C1CS(C2=CC(F)=C(SC3=NC(NC4=NNC(C)=C4)=C(OC)C(N5CCCCC5)=N3)C=C2)(=O)=O
Molekylvægt (g/mol) 631.67
Formel vægt 631.67
Procent renhed 95.03%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease