Learn More
Corning™ CellCube™ Modules, With Aseptic Connectors
BIOPHARMAGREENER_CHOICE
Provides a fast, simple, and compact method for the mass culture of attachment-dependent cells.
Brand:  Corning™ 3233
20268.00 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 17219109
  • 20409.88 DKK / Each
Only null left
Estimated Shipment: 18-12-2025
Description
  • Uses a tissue culture-treated growth surface for cell attachment, and continually perfuses the cells with fresh medium for increased cell productivity
  • The CellCube system provides an environment which more closely simulates in vivo conditions and reliably distributes nutrients and oxygen with low differential gradients across all cells within the modules.
  • The CellCube modules provide a traditional tissue culture-treated surface or Corning CellBIND™ surface for the growth of attachment dependent cells.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Growth Surface and Aseptic Connectors
1/Pk.
85,000 cm²
1/Cs.
Sterile
100-Layer
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Corning™ CellCube™ Modules, With Aseptic Connectors > 100