Få mere at vide
Cefuroxime sodium salt, 95%, Thermo Scientific™
1107.65 DKK - 4044.84 DKK
Kemiske identifikatorer
| CAS | 56238-63-2 |
|---|---|
| Molekylær formel | C16H15N4NaO8S |
| Molekylvægt (g/mol) | 446.366 |
| MDL nummer | MFCD09878727 |
| InChI nøgle | URDOHUPGIOGTKV-LNJPBTIASA-M |
| PubChem CID | 133687825 |
| IUPAC navn | sodium;(6S,7S)-3-(carbamoyloxymethyl)-7-[[(2Z)-2-(furan-2-yl)-2-methoxyiminoacetyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate |
| SMIL | CON=C(C1=CC=CO1)C(=O)NC2C3N(C2=O)C(=C(CS3)COC(=O)N)C(=O)[O-].[Na+] |
| Product Code | Brand | Mængde | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Mængde | Price | Quantity & Availability | |||||
|
15488589
|
Thermo Scientific Alfa Aesar
J66999.03 |
1g |
1231.36 DKK 1107.65 DKK / 1g Spare 123.71 DKK 10% Slukket |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
|
15498589
|
Thermo Scientific Alfa Aesar
J66999.06 |
5g |
4044.84 DKK
5g |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 56238-63-2 | |
| 446.366 | |
| URDOHUPGIOGTKV-LNJPBTIASA-M | |
| sodium;(6S,7S)-3-(carbamoyloxymethyl)-7-[[(2Z)-2-(furan-2-yl)-2-methoxyiminoacetyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate |
| C16H15N4NaO8S | |
| MFCD09878727 | |
| 133687825 | |
| CON=C(C1=CC=CO1)C(=O)NC2C3N(C2=O)C(=C(CS3)COC(=O)N)C(=O)[O-].[Na+] |
Specifications
| Cefuroxime sodium salt | |
| White to Yellow | |
| MFCD09878727 | |
| Freely soluble in water | |
| CON=C(C1=CC=CO1)C(=O)NC2C3N(C2=O)C(=C(CS3)COC(=O)N)C(=O)[O-].[Na+] | |
| 446.366 | |
| 446.37 |
| 56238-63-2 | |
| C16H15N4NaO8S | |
| 14,1954 | |
| URDOHUPGIOGTKV-LNJPBTIASA-M | |
| sodium;(6S,7S)-3-(carbamoyloxymethyl)-7-[[(2Z)-2-(furan-2-yl)-2-methoxyiminoacetyl]amino]-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate | |
| 133687825 | |
| 95% |
Safety and Handling
GHS H Statement
H334-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause an allergic skin reaction.
P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
H317-H334
missing translation for 'einecsNumber' : 260-073-1
missing translation for 'rtecsNumber' : XI0330000
missing translation for 'tsca' : No
missing translation for 'storageNote' : 2° to 8°C
Recommended Storage : Keep cold
- Cefuroxime sodium salt
Signal Word
- Fare
Hazard Category
- Respiratorisk sensibiliserende stof Kategori 1
- Hudsensibilisering Kategori 1
Hazard Statement
- H317-Kan forårsage allergisk hudreaktion.
- H334-Kan forårsage allergi- eller astmasymptomer eller åndedrætsbesvær ved indånding.
Precautionary Statement
- P280-Bær beskyttelseshandsker/beskyttelsestøj/øjenbeskyttelse/ ansigtsbeskyttelse.
- P284-[I tilfælde af utilstrækkelig ventilation], anvend åndedrætsværn.
- P302+P352-VED KONTAKT MED HUDEN: Vask med rigeligt vand/.
- P304+P340-VED INDÅNDING: Flyt personen til et sted med frisk luft og sørg for, at vejrtrækningen lettes.
- P333+P313-Ved hudirritation eller udslet: Søg lægehjælp.
- P342+P311-Ved luftvejssymptomer: Ring til en GIFTINFORMATION/læge/ .
Supplemental information
- MIXTURE LIST-Contain: Cefuroxime sodium salt
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.