Learn More
Cefotiam hydrochloride, MedChemExpress
MedChemExpress Cefotiam (SCE-963) hydrochloride is a parenteral cephalosporin antibiotic. Cefotiam hydrochloride has broad-spectrum activity against Gram-positive and Gram-negative bacteria.

523.71 DKK - 725.13 DKK
Specifications
Chemical Name or Material | Cefotiam hydrochloride |
---|---|
Physical Form | Solid |
CAS | 66309-69-1 |
Color | White |
Purity Grade Notes | Research |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16498588
![]() |
Medchem Express
HY-B0734A |
10 mg |
523.71 DKK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16414467
![]() |
Medchem Express
HY-B0734A |
50 mg |
725.13 DKK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cefotiam (SCE-963) hydrochloride is a parenteral cephalosporin antibiotic. Cefotiam hydrochloride has broad-spectrum activity against Gram-positive and Gram-negative bacteria.Specifications
Cefotiam hydrochloride | |
66309-69-1 | |
Research | |
Neuroscience-Neuromodulation | |
DMSO : 100 mg/mL (167.07 mM; Need ultrasonic) ∣H2O : 33.33 mg/mL (55.68 mM; Need ultrasonic) | |
598.55 | |
98.0% |
Solid | |
White | |
C18H25Cl2N9O4S3 | |
SCE-963 hydrochloride | |
O=C(C(N12)=C(CSC3=NN=NN3CCN(C)C)CS[C@]2([H])[C@H](NC(CC4=CSC(N)=N4)=O)C1=O)O.[H]Cl.[H]Cl | |
598.55 | |
Research |
Safety and Handling
missing translation for 'shelfLife' : 4°C, sealed storage, away from moisture∣The compound is unstable in solutions, freshly prepared is recommended.
Recommended Storage : 4°C, sealed storage, away from moisture∣The compound is unstable in solutions, freshly prepared is recommended.
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.