missing translation for 'onlineSavingsMsg'
Få mere at vide

CDKI-73, MedChemExpress

Artikelnummer. 16485168
Encompass_Preferred
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16485168 10 mg
16421397 50 mg
16417897 5 mg
16405876 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 16485168 Leverandør Medchem Express Leverandørnr. HY12445

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

MedChemExpress CDKI-73 (LS-007) is an orally active and highly efficacious CDK9 inhibitor, with Ki values of 4 nM, 4 nM and 3 nM for CDK9, CDK1 and CDK2, respectively. CDKI-73 down-regulates the RNAPII phosphorylation. CDKI-73 is also a novel pharmacological inhibitor of Rab11 cargo delivery and innate immune secretion.

CDKI-73 (LS-007) is an orally active and highly efficacious CDK9 inhibitor, with Ki values of 4 nM, 4 nM and 3 nM for CDK9, CDK1 and CDK2, respectively. CDKI-73 down-regulates the RNAPII phosphorylation. CDKI-73 is also a novel pharmacological inhibitor of Rab11 cargo delivery and innate immune secretion.

Tekniske data

Kemisk navn eller materiale CDKI-73
CAS 1421693-22-2
Farve Light Yellow
Renhedsgrad noter Research
Molekylær formel C15H15FN6O2S2
Mængde 10 mg
Synonym LS-007
Opløselighedsinformation DMSO : ≥ 52 mg/mL (131.83 mM)
SMIL CNC1=NC(C)=C(C2=C(F)C=NC(NC3=CC(S(=O)(N)=O)=CC=C3)=N2)S1
Molekylvægt (g/mol) 394.45
Formel vægt 394.45
Procent renhed 98.36%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-Kinase/protease
Vis mere Vis mindre

Research purposes only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.