missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
CD35 Mouse Monoclonal Antibody, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68033-1-IG-20UL
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
The CD35 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CD35. This antibody recognizes human antigen. The CD35 antibody has been validated for the following applications: IHC, IF-P, ELISA analysis.Tekniske data
| CD35 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Protein A purification | |
| RUO | |
| 1378 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Immunohistochemistry, Immunofluorescence, ELISA | |
| 1B8E6 | |
| Unconjugated | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human | |
| Monoclonal Antibody | |
| IgG1 |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion