missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Catalase Rabbit anti-Human, Mouse, Rat, Clone: S03-8E9, Novus Biologicals™
Rabbit Monoclonal Antibody
2185.00 DKK - 4415.00 DKK
Tekniske data
| Antigen | Catalase |
|---|---|
| Klon | S03-8E9 |
| Fortynding | Western Blot, ELISA, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 2.5 μg/mL, Immunofluorescence 2.5 μg/mL |
| Applikationer | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence |
| Klassifikation | Monoclonal |
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|
18367915
|
Novus Biologicals
NBP3-14914-25UL |
25 μg |
2185.00 DKK
25µL |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
|
18310356
|
Novus Biologicals
NBP3-14914-100UL |
100 μg |
4415.00 DKK
100µL |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
Beskrivelse
Catalase Monoclonal antibody specifically detects Catalase in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), ImmunofluorescenceTekniske data
| Catalase | |
| Western Blot, ELISA, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 2.5 μg/mL, Immunofluorescence 2.5 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| catalase, EC 1.11.1.6, MGC138422, MGC138424 | |
| Recombinant protein of mouse Catalase (Uniprot#: P24270) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| S03-8E9 | |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cellular Markers, Hypoxia, Lipid and Metabolism, Neuroscience, Peroxisome Markers, Signal Transduction | |
| 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
| 847 | |
| IgG | |
| Affinity-purified |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel