missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Catalase Rabbit anti-Human, Mouse, Rat, Clone: S03-8E9, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-14914-100UL
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Catalase Monoclonal antibody specifically detects Catalase in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence
Tekniske data
| Catalase | |
| Monoclonal | |
| Unconjugated | |
| 50nM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA with 0.01% Sodium Azide | |
| Rabbit | |
| Affinity-purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence | |
| S03-8E9 | |
| Western Blot, ELISA, Immunohistochemistry 2.5 μg/mL, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 2.5 μg/mL, Immunofluorescence 2.5 μg/mL | |
| catalase, EC 1.11.1.6, MGC138422, MGC138424 | |
| Recombinant protein of mouse Catalase (Uniprot#: P24270) | |
| 100 μg | |
| Cancer, Cellular Markers, Hypoxia, Lipid and Metabolism, Neuroscience, Peroxisome Markers, Signal Transduction | |
| 847 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion