Få mere at vide
Carbosulfan, MedChemExpress
Artikelnummer. 16472296
Encompass_Preferred
Skift visning
Mængde:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16472296 500 mg
16428836 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16472296 Leverandør Medchem Express Leverandørnr. HYB2015
MedChemExpress Carbosulfan inhibited relatively potently CYP3A4 and moderately CYP1A1/2 and CYP2C19 in pooled HLM (human livers).
Carbosulfan inhibited relatively potently CYP3A4 and moderately CYP1A1/2 and CYP2C19 in pooled HLM (human livers).
Tekniske data
Kemisk navn eller materiale Carbosulfan
CAS 55285-14-8
Farve Tan
Renhedsgrad noter Research
Molekylær formel C20H32N2O3S
Mængde 500 mg
Opløselighedsinformation DMSO : ≥ 100 mg/mL (262.78 mM)
SMIL O=C(OC1=C(OC(C)(C)C2)C2=CC=C1)N(SN(CCCC)CCCC)C
Molekylvægt (g/mol) 380.54
Formel vægt 380.54
Procent renhed 98.0%
Grad Research
Fysisk form Liquid
Til brug med (applikation) COVID-19-immunoregulation