Få mere at vide
Swann-Morton™ Skalpelblade i kulstofstål
Make surgical cuts and repairs using Swann Morton™ Carbon Steel Scalpel Blades in the operating room.
Brand: Swann-Morton™ 0111
Denne vare kan ikke returneres.
Se returpolitik
Beskrivelse
Make surgical cuts and repairs using Swann Morton™ Carbon Steel Scalpel Blades in the operating room.
- Carbon steel to BS EN 27740
- Handles and blades sold separately
Tekniske data
Carbon Steel | |
Used for Making Long Incisions in General Surgery and also in Autopsy Procedures | |
35 mm | |
Single-use | |
10/Pk. |
Scalpel Blade | |
# 24 | |
The # 24 fits Handles # 4, # 4L, # 4 Graduated and # 6B | |
BS EN 7740 |
Korrektion af produktindhold
Produkttitel
Swann-Morton™ Skalpelblade i kulstofstål > Fits Model: No. 24; Length: 35mm
Ser du en mulighed for forbedring?Del en indholdskorrektion