Få mere at vide
Carbocisteine Sulfoxide (Mixture of diastereomers), TRC
Artikelnummer. 17987854
Skift visning
Mængde:
250 mg
2.5 g
Pakningsstørrelse:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
17987854 2.5 g 2.5g
17918624 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17987854 Leverandør Toronto Research Chemicals Leverandørnr. TRCC1787802.5G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 5439-87-2
Molekylær formel C5 H9 N O5 S
Molekylvægt (g/mol) 195.19
Synonym (S)-Carboxymethyl-L-cysteine (R/S)-Sulphoxide
IUPAC navn (2R)-2-amino-3-(carboxymethylsulfinyl)propanoic acid
SMIL N[C@@H](CS(=O)CC(=O)O)C(=O)O
Tekniske data
Kemisk navn eller materiale (S)-Carboxymethyl-L-cysteine (RS)-Sulfoxide
Mængde 2.5 g
InChI formel InChI=1S/C5H9NO5S/c6-3(5(9)10)1-12(11)2-4(7)8/h3H,1-2,6H2,(H,7,8)(H,9,10)/t3-,12?/m0/s1
Formel vægt 195.0201