CAS RN 5439-87-2
IUPAC Navn:
—
Synonymer:
—
Molekylvægt (g/mol):
—
Molekylformel:
—
InChi Key:
—
SMILES:
—
1
–
2
af
2
Resultater
1
(S)-Carboxymethyl-L-cysteine (R/S)-Sulphoxide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Carbocisteine Sulfoxide (Mixture of diastereomers), TRC
CAS: 5439-87-2 Molekylær formel: C5 H9 N O5 S Molekylvægt (g/mol): 195.19 Synonym: (S)-Carboxymethyl-L-cysteine (R/S)-Sulphoxide IUPAC navn: (2R)-2-amino-3-(carboxymethylsulfinyl)propanoic acid SMIL: N[C@@H](CS(=O)CC(=O)O)C(=O)O