missing translation for 'onlineSavingsMsg'
Learn More

Emballator Cans With Snap on Handle

Made of HDPE. Emballator Cans With Snap on Handle is available in sizes between 2-10L.

Brand:  Emballator 48C0500-500-117

Encompass

 View more versions of this product

Product Code. 11334405

  • 27.57 DKK / Each
Only null left
In Stock
Add to basket
This item is not returnable. View return policy

Description

Description

  • Available in sizes between 2 - 10L
  • Material: HDPE
  • Approved for food contact
Specifications

Specifications

Can, Snap Handle
Natural
190 mm
284 mm
210 g
5 L
HDPE
140 mm
50 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
Emballator Cans With Snap on Handle

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.