missing translation for 'onlineSavingsMsg'
Learn More

Calcium 3-methyl-2-oxopentanoate, TRC

Product Code. 30363865
Change view
Click to view available options
Mængde:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30363865 50 mg
30362399 250 mg
30364461 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30363865 Supplier Toronto Research Chemicals Supplier No. TRCC28004050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66872-75-1
Molekylær formel 2 C6 H9 O3 . Ca
Molekylvægt (g/mol) 298.35
Synonym Pentanoic acid, 3-methyl-2-oxo-, calcium salt (2:1), Pentanoic acid, 3-methyl-2-oxo-, calcium salt (9CI), Pentanoic acid, 3-methyl-2-oxo-, calcium salt, (±)-, Calcium 3-methyl-2-oxopentanoate, calcium bis(3-methyl-2-oxopentanoate), α-keto-Isoleucine calcium, 3-Methyl-2-oxopentanoic acid calcium salt, Calcium (±)-3-methyl-2-oxovalerate
IUPAC navn calcium;3-methyl-2-oxopentanoate
SMIL [Ca+2].CCC(C)C(=O)C(=O)[O-].CCC(C)C(=O)C(=O)[O-]

Specifications

Kemisk navn eller materiale 3-Methyl-2-oxovaleric acid calcium salt
Mængde 50 mg
InChI formel InChI=1S/2C6H10O3.Ca/c2*1-3-4(2)5(7)6(8)9;/h2*4H,3H2,1-2H3,(H,8,9);/q;;+2/p-2
Formel vægt 298.0729
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.