Få mere at vide
Bupivacaine hydrochloride monohydrate, 98+%, Thermo Scientific™
Inhibits TREK-1 channels and depolarizes the cell membrane
544.78 DKK - 6701.59 DKK
Kemiske identifikatorer
| CAS | 73360-54-0 |
|---|---|
| Molekylær formel | C18H31ClN2O2 |
| Molekylvægt (g/mol) | 342.908 |
| MDL nummer | MFCD00078956 |
| InChI nøgle | HUCIWBPMHXGLFM-UHFFFAOYSA-N |
| Synonym | bupivacaine hydrochloride monohydrate, lac-43, marcaine tn, +/--1-butyl-2',6'-pipecoloxylidide monohydrochloride, monohydrate, 1-butyl-n-2,6-dimethylphenyl piperidine-2-carboxamide hydrate hydrochloride, bupivacaine hydrate hydrochloride, bupivacaine hcl h2-o, 2-piperidinecarboxamide, 1-butyl-n-2,6-dimethylphenyl-, monohydrochloride, monohydrate, bupivacaine hydrochloride ep, 1-butyl-n-2,6-dimethylphenyl piperidine-2-carboxamide hydrochloride hydrate |
| PubChem CID | 5282419 |
| IUPAC navn | 1-butyl-N-(2,6-dimethylphenyl)piperidine-2-carboxamide;hydrate;hydrochloride |
| SMIL | CCCCN1CCCCC1C(=O)NC2=C(C=CC=C2C)C.O.Cl |
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkode | Brand | Mængde | Pris | Antal & tilgængelighed | |||||
|
15455139
|
Thermo Scientific Alfa Aesar
J62835.03 |
1g |
544.78 DKK
1g |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
|
15465139
|
Thermo Scientific Alfa Aesar
J62835.06 |
5g |
1567.19 DKK
5g |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
|
15475139
|
Thermo Scientific Alfa Aesar
J62835.14 |
25g |
6701.59 DKK
25g |
Venligst log indfor at købe denne vare. Brug for en webkonto? Register dig hos os i dag! | |||||
Beskrivelse
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
ApplicationsIt is employed as Na+ channel blocker, local anesthetic and cAMP production inhibitor. It acts as a surfactant molecule possessing both hydrophilic and lipophilic properties, adrenergic antagonist, EC 3.1.1.8 (cholinesterase) inhibitor and EC 3.6.3.8 (Ca(2+)-transporting ATPase) inhibitor.
Solubility
Soluble in water to 50 mM, in DMSO to 100 mM and in ethanol to 100 mM.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Kemiske identifikatorer
| 73360-54-0 | |
| 342.908 | |
| HUCIWBPMHXGLFM-UHFFFAOYSA-N | |
| 5282419 | |
| CCCCN1CCCCC1C(=O)NC2=C(C=CC=C2C)C.O.Cl |
| C18H31ClN2O2 | |
| MFCD00078956 | |
| bupivacaine hydrochloride monohydrate, lac-43, marcaine tn, +/--1-butyl-2',6'-pipecoloxylidide monohydrochloride, monohydrate, 1-butyl-n-2,6-dimethylphenyl piperidine-2-carboxamide hydrate hydrochloride, bupivacaine hydrate hydrochloride, bupivacaine hcl h2-o, 2-piperidinecarboxamide, 1-butyl-n-2,6-dimethylphenyl-, monohydrochloride, monohydrate, bupivacaine hydrochloride ep, 1-butyl-n-2,6-dimethylphenyl piperidine-2-carboxamide hydrochloride hydrate | |
| 1-butyl-N-(2,6-dimethylphenyl)piperidine-2-carboxamide;hydrate;hydrochloride |
Tekniske data
| Bupivacaine hydrochloride monohydrate | |
| C18H31ClN2O2 | |
| UN2811 | |
| bupivacaine hydrochloride monohydrate, lac-43, marcaine tn, +/--1-butyl-2',6'-pipecoloxylidide monohydrochloride, monohydrate, 1-butyl-n-2,6-dimethylphenyl piperidine-2-carboxamide hydrate hydrochloride, bupivacaine hydrate hydrochloride, bupivacaine hcl h2-o, 2-piperidinecarboxamide, 1-butyl-n-2,6-dimethylphenyl-, monohydrochloride, monohydrate, bupivacaine hydrochloride ep, 1-butyl-n-2,6-dimethylphenyl piperidine-2-carboxamide hydrochloride hydrate | |
| HUCIWBPMHXGLFM-UHFFFAOYSA-N | |
| 1-butyl-N-(2,6-dimethylphenyl)piperidine-2-carboxamide;hydrate;hydrochloride | |
| 5282419 | |
| ≥98% |
| 73360-54-0 | |
| MFCD00078956 | |
| 14,1495 | |
| Soluble in water to 50 mM,in DMSO to 100 mM and in ethanol to 100 mM. | |
| CCCCN1CCCCC1C(=O)NC2=C(C=CC=C2C)C.O.Cl | |
| 342.908 | |
| 342.9 |
Sikkerhed og håndtering
GHS H Statement
H300-H310-H330
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P330-P361-P363-P501c
H300+H310+H330
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 241-917-8
missing translation for 'rtecsNumber' : TK6125000
missing translation for 'tsca' : No
Anbefalet opbevaring : Ambient temperatures
RUO – Research Use Only
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.