Learn More
Cole-Parmer™ Bulkhead Union Compression Fitting
Offers very good chemical resistance. Ideal when working with any rigid tubing, including tubing made from fluoropolymers, other rigid plastics and metals
420.00 DKK - 635.00 DKK
Specifications
Product Type | Bulkhead Union Compression Fitting |
---|---|
Autoclavable | Autoclavable |
Material | Stainless Steel |
Min. Operating Temperature (English) | -40°F |
Min. Operating Temperature (Metric) | -40°C |
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Tubing Size | Price | Quantity & Availability | |||||
11730429
|
Cole-Parmer™
31406-18 |
1/4 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11845443
|
Cole-Parmer™
31406-12 |
1/8 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11875443
|
Cole-Parmer™
31406-14 |
3/8 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11885443
|
Cole-Parmer™
31406-15 |
1/2 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
- Very good chemical resistance.
- Rates to 220psi at 77°F (25°C).
- Usable from 32 to 212°F (0 to +100°C).
- Compression fittings work well with rigid tubing such as PTFE or polypropylene.
- A compression fitting has a nut that compresses rigid tubing against fitting body for high-pressure applications.
- Compression connector family is ideal when working with rigid tubing.
- When selecting your fittings, consider your tubing type, material compatibility and temperature and pressure conditions.
Specifications
Bulkhead Union Compression Fitting | |
Stainless Steel | |
-40°C | |
220 psi | |
93°C |
Autoclavable | |
-40°F | |
Autoclave, Ethylene Oxide or Gamma | |
200°F |