missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer solution SrCO3 for AAS 0.02mg/L HCl matrix, Fisher chemical™

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
1445.00 DKK - 3065.00 DKK
Specifications
Physical Form | Liquid |
---|---|
Concentration | 0.02 mg/L |
Type | Buffer Solution |
Solvent or Matrix | HCl |
Chemical Name or Material | SrCO3 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
12994545
![]() |
Fisher Chemical
F 002 100 |
100 mL | Bottle |
1445.00 DKK
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12904555
![]() |
Fisher Chemical
F 002 500 |
500 mL | Bottle |
3065.00 DKK
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Liquid | |
Buffer Solution | |
SrCO3 |
0.02 mg/L | |
HCl |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Buffer solution SrCO3 for AAS 0.02mg/L HCl matrix, Fisher chemical™