missing translation for 'onlineSavingsMsg'
Learn More
Buffer Solution pH 2.00 (Glycine), NIST Standard Solution Ready to Use for pH Measurement, Fisher Chemical™ Product Code.: 10703324
compliance-icons

Buffer Solution pH 2.00 (Glycine), NIST Standard Solution Ready to Use for pH Measurement, Fisher Chemical™

Product Code. p-7073110 Shop All Fisher Chemical Products
500 mL, 500mL
Click to view available options
Quantity:
500 mL
2.5 L
Unit Size:
2.5 L
500mL
This item is not returnable. View return policy

Product Code. 10703324

Brand: Fisher Chemical J/2800/08

Quality Analysis and Quality Control Testing
Use promo code 24140 at checkout
View Available Promotions
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Buffer Solution pH 2.00 (Glycine), NIST Standard Solution Ready to Use for pH Measurement

TRUSTED_SUSTAINABILITY

Chemical Identifiers

MDL Number 217672

Specifications

Color Colorless
pH 2
Physical Form Liquid
Quantity 500 mL
Packaging HDPE plastic bottle

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Product Code
Lot Number2539500 Certificate TypeCertificate of Analysis Date09/06/2025 Product CodeJ/2800C/90,J/2800C/05,J/2800/15,J/2800/17,J/2800/08
Lot Number2533993 Certificate TypeCertificate of Analysis Date24/05/2025 Product CodeJ/2800C/90,J/2800C/05,J/2800/15,J/2800/17,J/2800/08
Lot Number2532635 Certificate TypeCertificate of Analysis Date24/05/2025 Product CodeJ/2800C/90,J/2800C/05,J/2800/15,J/2800/17,J/2800/08
compliance-icons
Product Identifier
  • Buffer solution pH 2 (glycine) Standard Buffer Solution
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P234-Keep only in original container.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Buffer Solution pH 2.00 (Glycine), NIST Standard Solution Ready to Use for pH Measurement, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.