missing translation for 'onlineSavingsMsg'
Learn More
Buffer Concentrated Solution pH9.2 (Borate), NIST Standard for pH measurement, Fisher Chemical™ Product Code.: 10246531

Buffer Concentrated Solution pH9.2 (Borate), NIST Standard for pH measurement, Fisher Chemical™

Product Code. p-7073121 Shop All Fisher Chemical Products
1 Ampule, 1 ampoule
Click to view available options
Quantity:
1 Ampule
6 Ampules
Unit Size:
1 ampoule
6 ampoules
This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

Product Code. 10246531

Brand: Fisher Chemical J/2870C/05

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

Chemical Identifiers

MDL Number 163147

Specifications

Color Colorless
Physical Form Liquid
Quantity 1 Ampule
Packaging Ampoule

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Product Code
Lot Number2538882 Certificate TypeCertificate of Analysis Date24/05/2025 Product CodeJ/2870/08,J/2870/17,J/2870C/05,J/2870C/90,J/2870/21,J/2870/15,J/2870/24
Lot Number2531534 Certificate TypeCertificate of Analysis Date24/05/2025 Product CodeJ/2870/08,J/2870/17,J/2870C/05,J/2870C/90,J/2870/21,J/2870/15,J/2870/24
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Buffer Concentrated Solution pH9.2 (Borate), NIST Standard for pH measurement, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.