missing translation for 'onlineSavingsMsg'
Learn More

Bromide, Ion chromatography standard solution, Specpure™ Br∣- 1000μg/mL

1170.00 DKK - 2475.00 DKK

Chemical Identifiers

MDL Number MFCD00134172
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11303657
SDS View Documents
Thermo Scientific Alfa Aesar
035552.AE
100 mL
1170.00 DKK
100mL
Please to purchase this item. Need a web account? Register with us today!
11415020
SDS View Documents
Thermo Scientific Alfa Aesar
035552.AP
500 mL
2475.00 DKK
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

MFCD00134172

Specifications

Liquid
MFCD00134172
Specpure™
Bromide Ion chromatography standard solution, Br- 1000μg/mL
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bromide, Ion chromatography standard solution, Specpure™ Br∣- 1000μg/mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.