missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bromfenac sodium, Thermo Scientific™
1110.00 DKK - 3700.00 DKK
Chemical Identifiers
CAS | 91714-93-1 |
---|---|
Molecular Formula | C15H11BrNNaO3 |
Molecular Weight (g/mol) | 356.15 |
MDL Number | MFCD03701673 |
InChI Key | HZFGMQJYAFHESD-UHFFFAOYSA-M |
IUPAC Name | sodium 2-[2-amino-3-(4-bromobenzoyl)phenyl]acetate |
SMILES | [Na+].NC1=C(CC([O-])=O)C=CC=C1C(=O)C1=CC=C(Br)C=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17535565
|
Thermo Scientific Acros
468922500 |
250 mg |
1110.00 DKK
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17525565
|
Thermo Scientific Acros
468920010 |
1 g |
3700.00 DKK
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
91714-93-1 | |
356.15 | |
HZFGMQJYAFHESD-UHFFFAOYSA-M | |
[Na+].NC1=C(CC([O-])=O)C=CC=C1C(=O)C1=CC=C(Br)C=C1 |
C15H11BrNNaO3 | |
MFCD03701673 | |
sodium 2-[2-amino-3-(4-bromobenzoyl)phenyl]acetate |
Specifications
Glass Bottle | |
Available | |
C15H11BrNNaO3 | |
Yellow to Orange | |
[Na+].NC1=C(CC([O-])=O)C=CC=C1C(=O)C1=CC=C(Br)C=C1 | |
356.15 |
91714-93-1 | |
Powder | |
MFCD03701673 | |
HZFGMQJYAFHESD-UHFFFAOYSA-M | |
sodium 2-[2-amino-3-(4-bromobenzoyl)phenyl]acetate | |
Bromfenac sodium |
Safety and Handling
Environment
Recommended Storage : Normal conditions
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Bromfenac sodium, Thermo Scientific™